Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163144.1 | NP_001156616.1 | 5 - 6 | 1116 | 94 | |
NM_001190483.1 | NP_001177412.1 | 5 - 6 | 1116 | 94 | ||
GenBank mRNA | AK086210.1 | - | 5 - 6 | 1103 | 95 | |
AK087337.1 | - | 5 - 6 | 1117 | 94 | ||
AK143998.1 | - | 5 - 6 | 1117 | 94 | ||
BC013068.1 | - | 5 - 6 | 736 | 94 | ||
D12619.1 | - | 5 - 6 | 705 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |