Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198019.2 | NP_932136.2 | 13 - 14 | 1748 | 118 | |
GenBank mRNA | AK050870.1 | - | 12 - 13 | 1874 | 118 | |
AK160295.1 | - | 5 - 6 | 900 | 118 | ||
BC070400.1 | - | 13 - 14 | 1748 | 5694341 | 118 | |
BC071270.1 | - | 13 - 14 | 1730 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |