Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011639.3 | NP_035769.1 | 2 - 3 | 410 | 71 | |
GenBank mRNA | AF097511.1 | - | 2 - 3 | 303 | 71 | |
AF116823.1 | - | 2 - 3 | 405 | 71 | ||
AK010435.1 | - | 2 - 3 | 411 | 71 | ||
AK146823.1 | - | 2 - 3 | 359 | 71 | ||
AK159387.1 | - | 2 - 3 | 379 | 71 | ||
AK166024.1 | - | 2 - 3 | 574 | 71 | ||
BC138685.1 | - | 2 - 3 | 342 | 8861706 | 71 | |
BC138686.1 | - | 2 - 3 | 342 | 8861707 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |