Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10526938 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526932 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526934 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526939 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526935 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526937 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526933 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526940 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10526936 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1441341_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10526931 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0500015430.mm.1 |
TC0500001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015430.mm.1 |
NM_001160265.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015430.mm.1 |
NM_001160265.1 |
Refseq |