Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500013397.mm.1 |
TC0500001487.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013397.mm.1 |
NM_177005.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013397.mm.1 |
NM_177005.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013392.mm.1 |
TC0500001487.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013392.mm.1 |
NM_177005.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013392.mm.1 |
NM_177005.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013393.mm.1 |
TC0500001487.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013393.mm.1 |
NM_177005.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013393.mm.1 |
NM_177005.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013395.mm.1 |
TC0500001487.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013395.mm.1 |
NM_177005.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013395.mm.1 |
NM_177005.4 |
Refseq |