Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184981.1 | NP_001171910.1 | 1 - 2 | 85 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Probe spans exons |