Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013833.3 | NP_001013855.1 | 5 - 6 | 927 | 92 | |
NM_011160.3 | NP_035290.1 | 5 - 6 | 850 | 92 | ||
GenBank mRNA | AF084547.1 | - | 5 - 6 | 850 | 92 | |
AK083991.1 | - | 5 - 6 | 927 | 92 | ||
BC113162.1 | - | 5 - 6 | 832 | 40060049 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |