Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010776.1 | NP_034906.1 | 2 - 3 | 173 | 75 | |
GenBank mRNA | AK149484.1 | - | 2 - 3 | 203 | 75 | |
BC010760.1 | - | 2 - 3 | 188 | 4212216 | 75 | |
D11440.1 | - | 2 - 3 | 173 | 75 | ||
S42294.1 | - | 2 - 3 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |