Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015876.2 | NP_001015876.1 | 12 - 13 | 1683 | 75 | |
GenBank mRNA | AK082884.1 | - | 12 - 13 | 1674 | 75 | |
BC027065.1 | - | 7 - 8 | 757 | 75 | ||
BC039634.1 | - | 1 - 2 | 107 | 75 | ||
BC068126.1 | - | 12 - 13 | 1591 | 6829456 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |