Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 10 - 11 | 1270 | 59 | |
GenBank mRNA | AJ005678.1 | - | 10 - 11 | 1228 | 59 | |
BC060184.1 | - | 10 - 11 | 1791 | 59 | ||
BC139010.1 | - | 10 - 11 | 1200 | 8862032 | 59 | |
BC139011.1 | - | 10 - 11 | 1200 | 8862033 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |