Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 2 - 3 | 314 | 83 | |
GenBank mRNA | AJ005678.1 | - | 2 - 3 | 272 | 83 | |
AK145898.1 | - | 2 - 3 | 300 | 83 | ||
AK158240.1 | - | 2 - 3 | 210 | 83 | ||
BC060184.1 | - | 2 - 3 | 314 | 83 | ||
BC139010.1 | - | 2 - 3 | 244 | 8862032 | 83 | |
BC139011.1 | - | 2 - 3 | 244 | 8862033 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |