Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207213.2 | NP_997096.2 | 9 - 10 | 1480 | 146 | |
GenBank mRNA | AY601646.1 | - | 8 - 9 | 1699 | 146 | |
BC054725.1 | - | 4 - 5 | 736 | 5699137 | 146 | |
BC057063.1 | - | 8 - 9 | 1228 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |