Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029569.3 | NP_083845.1 | 3 - 4 | 501 | 69 | |
GenBank mRNA | AF398966.1 | - | 3 - 4 | 412 | 69 | |
AK003770.1 | - | 3 - 4 | 502 | 69 | ||
BC108405.1 | - | 3 - 4 | 416 | 30288598 | 69 | |
BC120851.1 | - | 3 - 4 | 467 | 40129774 | 69 | |
BC132007.1 | - | 3 - 4 | 467 | 40130284 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |