Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011714.2 | NP_035844.2 | 8 - 9 | 3068 | 90 | |
GenBank mRNA | AF084480.1 | - | 8 - 9 | 3027 | 90 | |
AK037737.1 | - | 2 - 3 | 276 | 90 | ||
BC141399.1 | - | 8 - 9 | 2971 | 9056012 | 90 | |
BC171975.1 | - | 8 - 9 | 2971 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |