Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027891.4 | NP_082167.2 | 9 - 10 | 1353 | 58 | |
GenBank mRNA | AK004717.1 | - | 7 - 8 | 2315 | 58 | |
AK051910.1 | - | 9 - 10 | 1301 | 58 | ||
AK084948.1 | - | 9 - 10 | 1353 | 58 | ||
AK153976.1 | - | 9 - 10 | 1324 | 58 | ||
AK154352.1 | - | 9 - 10 | 1295 | 58 | ||
AK167020.1 | - | 9 - 10 | 1302 | 58 | ||
AK172708.1 | - | 9 - 10 | 1321 | 58 | ||
AK178976.1 | - | 1 - 2 | 102 | 58 | ||
AK187635.1 | - | 1 - 2 | 101 | 58 | ||
AK207344.1 | - | 1 - 2 | 60 | 58 | ||
AK218154.1 | - | 1 - 2 | 213 | 58 | ||
BC048397.1 | - | 9 - 10 | 1332 | 6396815 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |