Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009580.2 | NP_033606.2 | 7 - 8 | 1241 | 92 | |
GenBank mRNA | BC100347.1 | - | 7 - 8 | 1259 | 92 | |
BC125613.1 | - | 7 - 8 | 1241 | 40130128 | 92 | |
BC125615.1 | - | 7 - 8 | 1241 | 40130130 | 92 | |
U20448.1 | - | 7 - 8 | 1272 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |