Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021535.4 | NP_067510.3 | 2 - 3 | 313 | 115 | |
GenBank mRNA | AB044414.1 | - | 2 - 3 | 242 | 115 | |
AK003493.1 | - | 2 - 3 | 300 | 115 | ||
AK011140.1 | - | 2 - 3 | 312 | 115 | ||
AK019174.1 | - | 2 - 3 | 272 | 115 | ||
AK145982.1 | - | 2 - 3 | 312 | 115 | ||
AK159635.1 | - | 2 - 3 | 300 | 115 | ||
AK168813.1 | - | 2 - 3 | 307 | 115 | ||
BC020034.1 | - | 2 - 3 | 255 | 115 | ||
BC057446.1 | - | 2 - 3 | 274 | 3155760 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |