Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025444.3 | NP_001020615.1 | 6 - 7 | 811 | 63 | |
NM_001025445.2 | NP_001020616.1 | 6 - 7 | 806 | 63 | ||
NM_025545.4 | NP_079821.3 | 6 - 7 | 946 | 63 | ||
GenBank mRNA | AK005286.1 | - | 5 - 6 | 763 | 63 | |
AK010516.1 | - | 6 - 7 | 793 | 63 | ||
AK017522.1 | - | 5 - 6 | 774 | 63 | ||
AK077351.1 | - | 6 - 7 | 947 | 63 | ||
AK088928.1 | - | 5 - 6 | 496 | 63 | ||
AY040780.1 | - | 6 - 7 | 957 | 63 | ||
AY208844.1 | - | 6 - 7 | 958 | 63 | ||
BC021872.1 | - | 6 - 7 | 804 | 3994375 | 63 | |
BC068309.1 | - | 5 - 6 | 719 | 63 | ||
BC071213.1 | - | 6 - 7 | 798 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |