Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172711.2 | NP_766299.1 | 14 - 15 | 1770 | 81 | |
GenBank mRNA | AK081776.1 | - | 1 - 2 | 190 | 81 | |
AK084627.1 | - | 14 - 15 | 1751 | 81 | ||
AK160956.1 | - | 12 - 13 | 2783 | 81 | ||
AK162869.1 | - | 1770 | 81 | |||
BC034252.1 | - | 241 | 81 | |||
BC039660.1 | - | 7 - 8 | 854 | 81 | ||
BC057962.1 | - | 12 - 13 | 1460 | 5353891 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |