Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172711.2 | NP_766299.1 | 8 - 9 | 1004 | 152 | |
GenBank mRNA | AK053420.1 | - | 8 - 9 | 978 | 152 | |
AK084627.1 | - | 8 - 9 | 985 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |