Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039075.2 | NP_001034164.1 | 9 - 10 | 877 | 69 | |
NM_001039076.2 | NP_001034165.1 | 7 - 8 | 994 | 69 | ||
GenBank mRNA | AF114379.2 | - | 9 - 10 | 841 | 69 | |
AJ005621.1 | - | 9 - 10 | 870 | 69 | ||
AK004020.1 | - | 9 - 10 | 885 | 69 | ||
AK137181.1 | - | 7 - 8 | 977 | 69 | ||
AY206011.1 | - | 6 - 7 | 911 | 69 | ||
BC099596.1 | - | 9 - 10 | 844 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |