Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175174.4 | NP_780383.2 | 8 - 9 | 1852 | 114 | |
GenBank mRNA | AK004982.1 | - | 8 - 9 | 1589 | 114 | |
AK220195.1 | - | 8 - 9 | 1598 | 114 | ||
BC026739.1 | - | 3 - 4 | 561 | 114 | ||
BC057606.1 | - | 8 - 9 | 1852 | 6402367 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |