Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300017115.mm.1 |
NM_001170985.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017115.mm.1 |
NM_001170985.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017115.mm.1 |
NM_130867.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017115.mm.1 |
NM_130867.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017115.mm.1 |
TC0300002279.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017116.mm.1 |
NM_001170985.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017116.mm.1 |
NM_001170985.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017116.mm.1 |
NM_130867.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017116.mm.1 |
NM_130867.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017116.mm.1 |
TC0300002279.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017114.mm.1 |
NM_001170985.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017114.mm.1 |
NM_001170985.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017114.mm.1 |
NM_130867.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017114.mm.1 |
NM_130867.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017114.mm.1 |
TC0300002279.mm.1 |
MTA TC |