Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025642.1 | NP_079918.1 | 1 - 2 | 348 | 65 | |
GenBank mRNA | AK012533.1 | - | 1 - 2 | 349 | 65 | |
AK014084.1 | - | 1 - 2 | 317 | 65 | ||
AK163310.1 | - | 1 - 2 | 382 | 65 | ||
BC079900.1 | - | 1 - 2 | 360 | 30602551 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |