Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008133.4 | NP_032159.1 | 1 - 2 | 615 | 103 | |
GenBank mRNA | AK075971.1 | - | 1 - 2 | 519 | 103 | |
AK089152.1 | - | 1 - 2 | 518 | 103 | ||
AK150637.1 | - | 1 - 2 | 615 | 103 | ||
BC052724.1 | - | 1 - 2 | 589 | 6812055 | 103 | |
BC057347.1 | - | 1 - 2 | 589 | 6812055 | 103 | |
X57024.1 | - | 1 - 2 | 520 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |