Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400013406.mm.1 |
NM_029389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013406.mm.1 |
NM_029389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013406.mm.1 |
TC1400001723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013408.mm.1 |
NM_029389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013408.mm.1 |
NM_029389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013408.mm.1 |
TC1400001723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013404.mm.1 |
NM_029389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013404.mm.1 |
NM_029389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013404.mm.1 |
TC1400001723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013405.mm.1 |
NM_029389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013405.mm.1 |
NM_029389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013405.mm.1 |
TC1400001723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013403.mm.1 |
NM_029389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013403.mm.1 |
NM_029389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013403.mm.1 |
TC1400001723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013407.mm.1 |
NM_029389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013407.mm.1 |
NM_029389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013407.mm.1 |
TC1400001723.mm.1 |
MTA TC |