Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173012.3 | NP_766600.2 | 5 - 6 | 1184 | 62 | |
GenBank mRNA | AK031563.1 | - | 5 - 6 | 1037 | 62 | |
AK163507.1 | - | 5 - 6 | 1038 | 62 | ||
BC050979.1 | - | 1 - 2 | 89 | 62 | ||
BC055685.1 | - | 5 - 6 | 1184 | 6403034 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |