Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900011162.mm.1 |
NM_017375.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011162.mm.1 |
NM_017375.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011162.mm.1 |
TC1900001227.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011163.mm.1 |
NM_017375.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011163.mm.1 |
NM_017375.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011163.mm.1 |
TC1900001227.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011164.mm.1 |
NM_017375.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011164.mm.1 |
NM_017375.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011164.mm.1 |
TC1900001227.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011166.mm.1 |
NM_017375.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011166.mm.1 |
NM_017375.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011166.mm.1 |
TC1900001227.mm.1 |
MTA TC |