Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011960.2 | NP_036090.2 | 7 - 8 | 1982 | 67 | |
GenBank mRNA | AF079557.1 | - | 7 - 8 | 1731 | 67 | |
AK036656.1 | - | 7 - 8 | 2005 | 67 | ||
AK088708.1 | - | 4 - 5 | 337 | 67 | ||
AK160742.1 | - | 6 - 7 | 1456 | 67 | ||
AK163388.1 | - | 7 - 8 | 2009 | 67 | ||
AK164013.1 | - | 7 - 8 | 1969 | 67 | ||
BC050892.1 | - | 7 - 8 | 1981 | 67 | ||
BC053037.1 | - | 7 - 8 | 1965 | 67 | ||
BC059827.1 | - | 7 - 8 | 1991 | 67 | ||
EF382673.1 | - | 5 - 6 | 691 | 67 | ||
JQ890227.1 | - | 4 - 5 | 488 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |