Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289686.1 | NP_001276615.1 | 7 - 8 | 708 | 107 | |
NM_001289687.1 | NP_001276616.1 | 4 - 5 | 579 | 107 | ||
NM_001289688.1 | NP_001276617.1 | 4 - 5 | 753 | 107 | ||
NM_027871.2 | NP_082147.1 | 5 - 6 | 553 | 107 | ||
GenBank mRNA | AK004600.2 | - | 5 - 6 | 524 | 107 | |
AK029872.1 | - | 8 - 9 | 738 | 107 | ||
BC005517.1 | - | 4 - 5 | 550 | 3494309 | 107 | |
BC012262.1 | - | 4 - 5 | 531 | 4192378 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |