Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpte | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199257.2 | NP_954866.2 | 17 - 18 | 1469 | 102 | |
GenBank mRNA | AJ311311.1 | - | 17 - 18 | 1472 | 102 | |
AJ311312.1 | - | 17 - 18 | 1557 | 102 | ||
AJ311313.1 | - | 17 - 18 | 1472 | 102 | ||
BC107329.2 | - | 17 - 18 | 1264 | 40054415 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |