Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
TC0500000400.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177697.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177697.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177698.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177698.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177699.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177699.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177700.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_001177700.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_177678.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003999.mm.1 |
NM_177678.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
TC0500000400.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177697.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177697.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177698.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177698.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177699.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177699.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177700.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_001177700.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_177678.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004000.mm.1 |
NM_177678.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004001.mm.1 |
TC0500000400.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500004001.mm.1 |
NM_001177697.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004001.mm.1 |
NM_001177697.1 |
Refseq |