Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035239.2 | NP_001030316.1 | 3 - 4 | 468 | 75 | |
NM_001035240.2 | NP_001030317.1 | 3 - 4 | 468 | 75 | ||
NM_001035241.2 | NP_001030318.1 | 3 - 4 | 468 | 75 | ||
NM_001035242.1 | NP_001030319.1 | 3 - 4 | 468 | 75 | ||
NM_001035243.2 | NP_001030320.1 | 3 - 4 | 468 | 75 | ||
NM_001035244.1 | NP_001030321.1 | 3 - 4 | 516 | 75 | ||
NM_001035245.1 | NP_001030322.1 | 3 - 4 | 516 | 75 | ||
NM_001035246.1 | NP_001030323.1 | 3 - 4 | 516 | 75 | ||
NM_177341.4 | NP_796315.2 | 3 - 4 | 535 | 75 | ||
GenBank mRNA | AJ544532.1 | - | 3 - 4 | 468 | 75 | |
AJ544533.1 | - | 3 - 4 | 468 | 75 | ||
AJ544534.1 | - | 3 - 4 | 468 | 75 | ||
AJ544535.1 | - | 3 - 4 | 468 | 75 | ||
AJ544536.1 | - | 3 - 4 | 468 | 75 | ||
AK044454.1 | - | 3 - 4 | 537 | 75 | ||
AK044533.1 | - | 3 - 4 | 314 | 75 | ||
AK046908.1 | - | 3 - 4 | 349 | 75 | ||
AK051934.1 | - | 3 - 4 | 314 | 75 | ||
AK052064.1 | - | 3 - 4 | 322 | 75 | ||
AK053673.1 | - | 3 - 4 | 353 | 75 | ||
AK077331.1 | - | 3 - 4 | 348 | 75 | ||
AK082141.1 | - | 3 - 4 | 350 | 75 | ||
AK084402.1 | - | 3 - 4 | 321 | 75 | ||
AK085182.1 | - | 3 - 4 | 516 | 75 | ||
AK142504.1 | - | 3 - 4 | 349 | 75 | ||
AK161909.1 | - | 3 - 4 | 349 | 75 | ||
AK163890.1 | - | 3 - 4 | 343 | 75 | ||
AK173218.1 | - | 3 - 4 | 539 | 75 | ||
BC037449.1 | - | 3 - 4 | 364 | 75 | ||
BC141400.1 | - | 3 - 4 | 511 | 9056013 | 75 | |
BC141412.1 | - | 3 - 4 | 511 | 9056025 | 75 | |
HM064407.1 | - | 3 - 4 | 560 | 75 | ||
HM064408.1 | - | 3 - 4 | 560 | 75 | ||
HM064409.1 | - | 3 - 4 | 560 | 75 | ||
HM064410.1 | - | 3 - 4 | 560 | 75 | ||
HM064411.1 | - | 3 - 4 | 560 | 75 | ||
JF706722.1 | - | 3 - 4 | 531 | 75 | ||
JX644970.1 | - | 3 - 4 | 560 | 75 | ||
JX644971.1 | - | 3 - 4 | 560 | 75 | ||
JX644972.1 | - | 3 - 4 | 560 | 75 | ||
JX644973.1 | - | 3 - 4 | 560 | 75 | ||
JX644974.1 | - | 3 - 4 | 560 | 75 | ||
JX644975.1 | - | 3 - 4 | 560 | 75 | ||
JX644976.1 | - | 3 - 4 | 560 | 75 | ||
JX644977.1 | - | 3 - 4 | 560 | 75 | ||
JX644978.1 | - | 3 - 4 | 560 | 75 | ||
JX644979.1 | - | 3 - 4 | 560 | 75 | ||
JX644980.1 | - | 3 - 4 | 560 | 75 | ||
JX644981.1 | - | 3 - 4 | 560 | 75 | ||
JX644982.1 | - | 3 - 4 | 560 | 75 | ||
JX644983.1 | - | 3 - 4 | 560 | 75 | ||
JX644984.1 | - | 3 - 4 | 560 | 75 | ||
JX644985.1 | - | 3 - 4 | 560 | 75 | ||
JX644986.1 | - | 3 - 4 | 560 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |