Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK132980.1 | - | 8 - 9 | 1292 | 122 | |
AV269315.2 | - | 2 - 3 | 239 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |