Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076605.1 | - | 9 - 10 | 1144 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |