Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145221.2 | NP_660256.1 | 5 - 6 | 464 | 98 | |
GenBank mRNA | AK030445.1 | - | 5 - 6 | 344 | 98 | |
AK033566.1 | - | 5 - 6 | 435 | 98 | ||
AK045438.1 | - | 5 - 6 | 400 | 98 | ||
AK049307.1 | - | 5 - 6 | 451 | 98 | ||
AK146356.1 | - | 5 - 6 | 435 | 98 | ||
AY113705.1 | - | 5 - 6 | 429 | 98 | ||
BC063751.1 | - | 5 - 6 | 476 | 6511401 | 98 | |
BU514763.1 | - | 5 - 6 | 475 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |