Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144500.3 | NP_653083.2 | 9 - 10 | 1077 | 96 | |
GenBank mRNA | AK048537.1 | - | 9 - 10 | 997 | 96 | |
AK145292.1 | - | 9 - 10 | 1109 | 96 | ||
AK161695.1 | - | 9 - 10 | 1016 | 96 | ||
AK166059.1 | - | 9 - 10 | 1056 | 96 | ||
AK170306.1 | - | 9 - 10 | 1061 | 96 | ||
AK170669.1 | - | 9 - 10 | 1078 | 96 | ||
AK182769.1 | - | 3 - 4 | 265 | 96 | ||
AK220257.1 | - | 3 - 4 | 396 | 96 | ||
BC026804.1 | - | 9 - 10 | 1084 | 4159217 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |