Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 24 - 25 | 3502 | 136 | |
GenBank mRNA | AF057367.1 | - | 24 - 25 | 3482 | 136 | |
AK142512.1 | - | 24 - 25 | 3418 | 136 | ||
AK162410.1 | - | 22 - 23 | 3075 | 136 | ||
AK164917.1 | - | 11 - 12 | 1947 | 136 | ||
AK220463.1 | - | 25 - 26 | 3459 | 136 | ||
BC053061.1 | - | 24 - 25 | 3484 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |