Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013927.2 | NP_038955.1 | 5 - 6 | 705 | 90 | |
GenBank mRNA | AJ243572.1 | - | 5 - 6 | 705 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mus musculus Probe spans exons |