Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133907.3 | NP_598668.1 | 8 - 9 | 1266 | 85 | |
GenBank mRNA | AK029973.1 | - | 8 - 9 | 1076 | 85 | |
AK045211.1 | - | 8 - 9 | 1064 | 85 | ||
AK046056.1 | - | 8 - 9 | 1267 | 85 | ||
AK122187.1 | - | 8 - 9 | 1201 | 85 | ||
BC059245.1 | - | 8 - 9 | 1081 | 85 | ||
BC120731.1 | - | 8 - 9 | 1013 | 40129654 | 85 | |
BC137626.1 | - | 8 - 9 | 1013 | 8860645 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |