Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033457.2 | NP_001028629.2 | 3 - 4 | 1304 | 90 | |
GenBank mRNA | AK148406.1 | - | 3 - 4 | 1308 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |