Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF025653.1 | - | 2 - 3 | 202 | 154 | |
AF034568.1 | - | 2 - 3 | 256 | 154 | ||
AK002922.1 | - | 2 - 3 | 305 | 154 | ||
AK079488.1 | - | 2 - 3 | 302 | 154 | ||
AK142016.1 | - | 2 - 3 | 299 | 154 | ||
AK150497.1 | - | 2 - 3 | 309 | 154 | ||
AK151463.1 | - | 2 - 3 | 323 | 154 | ||
AK160007.1 | - | 2 - 3 | 311 | 154 | ||
AK160706.1 | - | 2 - 3 | 289 | 154 | ||
AK169305.1 | - | 2 - 3 | 275 | 154 | ||
BC043657.1 | - | 2 - 3 | 274 | 5343539 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |