Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024241.2 | NP_077203.2 | 9 - 10 | 1662 | 87 | |
GenBank mRNA | AK050683.1 | - | 9 - 10 | 1648 | 87 | |
AK084967.1 | - | 6 - 7 | 1261 | 87 | ||
BC067395.1 | - | 9 - 10 | 1654 | 30460089 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |