Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tapt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173764.3 | NP_776125.2 | 12 - 13 | 1306 | 78 | |
GenBank mRNA | AK030002.1 | - | 7 - 8 | 548 | 78 | |
BC096038.1 | - | 9 - 10 | 785 | 78 | ||
BC110670.1 | - | 11 - 12 | 1027 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |