Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tapt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173764.3 | NP_776125.2 | 1 - 2 | 193 | 82 | |
GenBank mRNA | CN459607.1 | - | 1 - 2 | 193 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |