Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177850.1 | NP_001171321.1 | 10 - 11 | 1058 | 69 | |
NM_001177852.1 | NP_001171323.1 | 10 - 11 | 859 | 69 | ||
NM_001177853.1 | NP_001171324.1 | 10 - 11 | 826 | 69 | ||
NM_001177854.1 | NP_001171325.1 | 8 - 9 | 688 | 69 | ||
NM_023066.3 | NP_075553.2 | 10 - 11 | 1058 | 69 | ||
GenBank mRNA | AK030293.1 | - | 10 - 11 | 982 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |