Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153798.2 | NP_722493.2 | 10 - 11 | 1528 | 84 | |
GenBank mRNA | BC016415.1 | - | 10 - 11 | 1514 | 84 | |
BC038472.1 | - | 10 - 11 | 1521 | 5026936 | 84 | |
CN690754.1 | - | 1 - 2 | 52 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |