Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177815.3 | NP_808483.2 | 9 - 10 | 993 | 90 | |
GenBank mRNA | BC082539.1 | - | 9 - 10 | 968 | 90 | |
BC094336.1 | - | 9 - 10 | 961 | 90 | ||
BC141275.1 | - | 9 - 10 | 987 | 9055887 | 90 | |
BC145606.1 | - | 9 - 10 | 900 | 9054172 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |