Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp10d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153389.3 | NP_700438.3 | 6 - 7 | 1142 | 62 | |
NR_003966.1 | 6 - 7 | 1156 | 62 | |||
GenBank mRNA | AJ441079.1 | - | 6 - 7 | 1142 | 62 | |
AK036646.1 | - | 5 - 6 | 807 | 62 | ||
BC141328.1 | - | 6 - 7 | 993 | 9055940 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |