Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010568.2 | NP_034698.2 | 14 - 15 | 3409 | 85 | |
GenBank mRNA | J05149.1 | - | 14 - 15 | 2968 | 85 | |
KC356871.1 | - | 15 - 16 | 2956 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mus musculus Probe spans exons |